Assay Details
Target Gene Details
Entrez Gene ID: | 2274 |
Gene Name: | four and a half LIM domains 2 |
Gene Aliases: |
AAG11, DRAL, FHL-2, SLIM-3, SLIM3 |
Location: |
Chr.2:105360826-105438773 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL2 | NM_001039492.2 | NP_001034581.1 | ||
| NM_001318894.1 | NP_001305823.1 | |||
| NM_001318895.1 | NP_001305824.1 | |||
| NM_001318896.1 | NP_001305825.1 | |||
| NM_001318897.1 | NP_001305826.1 | |||
| NM_001318898.1 | NP_001305827.1 | |||
| NM_001318899.1 | NP_001305828.1 | |||
| NM_001450.3 | NP_001441.4 | |||
| NM_201555.1 | NP_963849.1 | |||
| NM_201557.3 | NP_963851.2 | |||
| XM_011510798.2 | XP_011509100.1 | |||
| AB158503.1 | BAD69710.1 | |||
| AK313841.1 | ||||
| AL521653.3 | ||||
| AY634685.1 | AAV35468.1 | |||
| BC012742.2 | AAH12742.1 | |||
| BC014397.1 | AAH14397.1 | |||
| BC093049.1 | ||||
| BI562043.1 | ||||
| BM559077.1 | ||||
| BT006960.1 | AAP35606.1 | |||
| CF552311.1 | ||||
| CR457437.1 | CAG33718.1 | |||
| CR936682.1 | ||||
| CR936799.1 | 3 | 2044 | ||
| DQ266432.1 | ABB73038.1 | |||
| DQ307067.1 | ABC25549.1 | |||
| JQ313902.1 | ||||
| L42176.1 | AAA85333.1 | |||
| U29332.1 | AAC52073.1 |
More Information
Set Membership: |
|

Genomic Map