This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04682866_cn
See other HSPD1 CNV Assays ›
Gene Symbol
HSPD1
Assay Reference Genome
Location

Chr.2:197488137 on build GRCh38
Cytoband
2q33.1
Assay ID Hs04682866_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

3329

Gene Name:

heat shock protein family D (Hsp60) member 1

Gene Aliases:

CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13

Location:

Chr.2:197486584-197500274 on Build GRCh38

Assay Gene Location:

Within Intron 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HSPD1 NM_002156.4 NP_002147.2
NM_199440.1 NP_955472.1
AK298368.1
AK298632.1
AK298844.1
AK312240.1
BC002676.2 AAH02676.1
BC003030.1 AAH03030.1
BC067082.1 AAH67082.1
BC073746.1 AAH73746.1
BX647145.1
EU719626.1
KU178139.1
KU178140.1
KU178141.1
M22382.1 AAA60127.1
M34664.1 AAA36022.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1014881 Chr.2:197432432 - 197668882 on Build GRCh38 Gain MOB4 HSPE1 HSPD1 SNORA105B RFTN2 COQ10B HSPE1-MOB4 SF3B1
nsv963757 Chr.2:197486354 - 197489175 on Build GRCh38 Duplication HSPD1 SNORA105B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

B cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
positive regulation of T cell mediated immune response to tumor cell
'de novo' protein folding
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to unfolded protein
response to cold
viral process
positive regulation of interferon-alpha production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
protein refolding
B cell proliferation
T cell activation
B cell activation
positive regulation of macrophage activation
positive regulation of apoptotic process
negative regulation of apoptotic process
isotype switching to IgG isotypes
protein stabilization
positive regulation of T cell activation
chaperone-mediated protein complex assembly
protein maturation
lipopolysaccharide binding
p53 binding
DNA replication origin binding
single-stranded DNA binding
double-stranded RNA binding
protein binding
ATP binding
ATPase activity
ubiquitin protein ligase binding
poly(A) RNA binding
unfolded protein binding
chaperone binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline