This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04722441_cn
See other CHL1 CNV Assays ›
Gene Symbol
CHL1
Assay Reference Genome
Location

Chr.3:197854 on build GRCh38
Cytoband
3p26.3
Assay ID Hs04722441_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10752

Gene Name:

cell adhesion molecule L1 like

Gene Aliases:

CALL, L1CAM2

Location:

Chr.3:196588-409417 on Build GRCh38

Assay Gene Location:

Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHL1 NM_001253387.1 NP_001240316.1
NM_006614.3 NP_006605.2
NR_045572.1 1 212
XM_006712938.1 XP_006713001.1
XM_006712939.3 XP_006713002.1
XM_006712940.3 1 152 XP_006713003.1
XM_011533292.1 1 211 XP_011531594.1
XM_011533294.1 XP_011531596.1
XM_011533295.1 1 211 XP_011531597.1
XM_011533296.1 XP_011531598.1
XM_017005566.1 XP_016861055.1
XM_017005567.1 XP_016861056.1
XM_017005569.1 XP_016861058.1
XM_017005570.1 1 152 XP_016861059.1
XM_017005571.1 XP_016861060.1
XM_017005572.1 XP_016861061.1
XM_017005573.1 XP_016861062.1
AB209329.1 BAD92566.1
AF002246.1 AAB60937.1
AK126878.1 1 211
DA778898.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1010970 Chr.3:188327 - 1623352 on Build GRCh38 Gain CHL1-AS1 CNTN6 CHL1 LOC101927174 LINC01266
esv2759122 Chr.3:29482 - 756820 on Build GRCh38 Gain+Loss CHL1-AS1 CHL1 LOC101927174 LINC01266
esv3594873 Chr.3:36658 - 319672 on Build GRCh38 Gain CHL1 LOC101927174
nsv469898 Chr.3:21733 - 886773 on Build GRCh38 Loss CHL1-AS1 CHL1 LOC101927174 LOC102723448 LINC01266
esv2667629 Chr.3:39576 - 1039187 on Build GRCh38 Deletion CHL1-AS1 CNTN6 CHL1 LOC101927174 LINC01266
nsv428412 Chr.3:29467 - 326726 on Build GRCh38 Gain CHL1 LOC101927174
nsv508206 Chr.3:183024 - 203282 on Build GRCh38 Deletion CHL1 LOC101927174
nsv1001867 Chr.3:193194 - 252963 on Build GRCh38 Gain CHL1 LOC101927174
nsv589247 Chr.3:181998 - 209030 on Build GRCh38 Loss CHL1 LOC101927174
nsv589283 Chr.3:192626 - 321571 on Build GRCh38 Gain CHL1 LOC101927174
dgv4607n100 Chr.3:18655 - 244340 on Build GRCh38 Gain CHL1 LOC101927174 LOC102723448
esv3893585 Chr.3:44574 - 267680 on Build GRCh38 Gain CHL1 LOC101927174
dgv4613n100 Chr.3:52474 - 1799230 on Build GRCh38 Loss CHL1-AS1 CNTN6 CHL1 LOC101927174 LINC01266
esv2751984 Chr.3:168062 - 253971 on Build GRCh38 Gain CHL1 LOC101927174
nsv3670 Chr.3:180797 - 207095 on Build GRCh38 Deletion CHL1 LOC101927174
nsv589246 Chr.3:143598 - 212269 on Build GRCh38 Gain CHL1 LOC101927174
nsv589242 Chr.3:47730 - 388999 on Build GRCh38 Gain CHL1-AS1 CHL1 LOC101927174

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
cell adhesion
signal transduction
axon guidance
adult locomotory behavior
axon regeneration
exploration behavior
negative regulation of neuron apoptotic process
cognition
protease binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-548c9458fd-h97b8:80/100.66.78.50:80.
git-commit: b50e517baec6e51dce58f17cb59caa961a96861c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.0-Offline