This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04786687_cn
See other SETMAR CNV Assays ›
Gene Symbol
SETMAR
Assay Reference Genome
Location

Chr.3:4305924 on build GRCh38
Cytoband
3p26.1
Assay ID Hs04786687_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

6419

Gene Name:

SET domain and mariner transposase fusion gene

Gene Aliases:

METNASE, Mar1

Location:

Chr.3:4303304-4320649 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SETMAR NM_001243723.1 NP_001230652.1
NM_001276325.1 NP_001263254.1
NM_001320676.1 NP_001307605.1
NM_001320677.1 NP_001307606.1
NM_001320678.1 NP_001307607.1
NM_006515.3 NP_006506.3
XM_006713292.3 XP_006713355.1
XM_006713294.3 XP_006713357.1
XM_017007032.1 XP_016862521.1
AF054989.1 AAC09350.1
AK222734.1 BAD96454.1
AK297865.1
AK302296.1
AK307537.1
AK311418.1
AY952295.1 AAY29570.1
BC008931.2 AAH08931.2
BC011635.2 AAH11635.1
BG479260.1
DC357038.1
U80776.1 AAC52012.1

Target Gene Details

Entrez Gene ID:

285362

Gene Name:

sulfatase modifying factor 1

Gene Aliases:

AAPA3037, FGE, UNQ3037

Location:

Chr.3:4034714-4467282 on Build GRCh38

Assay Gene Location:

Within Intron 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SUMF1 XM_011533624.2 XP_011531926.1
XM_011533625.2 XP_011531927.1
XM_017006252.1 XP_016861741.1
XM_017006253.1 XP_016861742.1
XM_017006254.1 XP_016861743.1
AY358092.1 AAQ88459.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3575535 Chr.3:4292219 - 4362841 on Build GRCh38 Gain SUMF1 SETMAR
dgv272n21 Chr.3:4080585 - 4319799 on Build GRCh38 Loss SUMF1 SETMAR
nsv1006124 Chr.3:4235682 - 4310067 on Build GRCh38 Loss SUMF1 SETMAR
esv3893640 Chr.3:4196512 - 4421324 on Build GRCh38 Loss SUMF1 SETMAR
esv2759126 Chr.3:4096685 - 4378673 on Build GRCh38 Loss SUMF1 SETMAR
nsv469899 Chr.3:4281224 - 4315301 on Build GRCh38 Loss SUMF1 SETMAR
nsv589451 Chr.3:4231805 - 4473507 on Build GRCh38 Loss SUMF1 SETMAR
nsv997886 Chr.3:4294748 - 4373592 on Build GRCh38 Gain SUMF1 SETMAR
nsv1003234 Chr.3:4298679 - 4344910 on Build GRCh38 Gain SUMF1 SETMAR

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

viral or transposable element protein histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

DNA double-strand break processing
DNA catabolic process, endonucleolytic
double-strand break repair via nonhomologous end joining
cell proliferation
histone H3-K36 methylation
DNA integration
replication fork processing
mitotic DNA integrity checkpoint
histone H3-K4 methylation
negative regulation of cell cycle arrest
nucleic acid phosphodiester bond hydrolysis
histone H3-K36 dimethylation
positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity
positive regulation of double-strand break repair via nonhomologous end joining
negative regulation of chromosome organization
glycosphingolipid metabolic process
post-translational protein modification
oxidation-reduction process
single-stranded DNA endodeoxyribonuclease activity
double-stranded DNA binding
single-stranded DNA binding
endonuclease activity
protein binding
zinc ion binding
histone methyltransferase activity (H3-K4 specific)
protein homodimerization activity
structure-specific DNA binding
DNA topoisomerase binding
histone methyltransferase activity (H3-K36 specific)
oxidoreductase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0