This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04795645_cn
See other SEPSECS CNV Assays ›
Gene Symbol
SEPSECS
Assay Reference Genome
Location

Chr.4:25133493 on build GRCh38
Cytoband
4p15.2
Assay ID Hs04795645_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51091

Gene Name:

Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase

Gene Aliases:

LP, PCH2D, SLA, SLA/LP

Location:

Chr.4:25120005-25160582 on Build GRCh38

Assay Gene Location:

Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SEPSECS NM_016955.3 NP_058651.3
XM_011513846.2 XP_011512148.1
XM_011513847.2 XP_011512149.1
XM_011513848.1 XP_011512150.1
XM_017008277.1 XP_016863766.1
XM_017008278.1 XP_016863767.1
AF146396.2 AAD33963.2
AF282065.1 AAG00491.1
AJ238617.1 CAB62209.1
AJ277541.1 CAB89517.1
AK292476.1
BC117202.1
BC126213.1
BX648976.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv4277 Chr.4:25127698 - 25172730 on Build GRCh38 Deletion SEPSECS-AS1 SEPSECS

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

selenocysteine incorporation
selenocysteine metabolic process
selenocysteinyl-tRNA(Sec) biosynthetic process
tRNA binding
protein binding
phosphoseryl-selenocysteinyl-tRNA selenium transferase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline