This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04829445_cn
See other EIF4E CNV Assays ›
Gene Symbol
EIF4E
Assay Reference Genome
Location

Chr.4:98888173 on build GRCh38
Cytoband
4q23
Assay ID Hs04829445_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

1977

Gene Name:

eukaryotic translation initiation factor 4E

Gene Aliases:

AUTS19, CBP, EIF4E1, EIF4EL1, EIF4F, eIF-4E

Location:

Chr.4:98878456-98930635 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4E NM_001130678.1 NP_001124150.1
NM_001130679.1 NP_001124151.1
NM_001968.3 NP_001959.1
XM_006714126.3 XP_006714189.1
AK295083.1
AK300982.1
AV708571.1
BC012611.1 AAH12611.1
BC035166.1 AAH35166.1
BC043226.1 AAH43226.1
BC107715.1 AAI07716.1
BM849222.1
DC388666.1
KJ534830.1
M15353.1 AAC13647.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
nuclear-transcribed mRNA poly(A) tail shortening
behavioral fear response
RNA export from nucleus
mRNA export from nucleus
translational initiation
regulation of translation
viral process
negative regulation of translation
stem cell population maintenance
lung development
negative regulation of neuron differentiation
positive regulation of mitotic cell cycle
RNA cap binding
translation initiation factor activity
protein binding
enzyme binding
eukaryotic initiation factor 4G binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline