This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04838569_cn
See other HADH CNV Assays ›
Gene Symbol
HADH
Assay Reference Genome
Location

Chr.4:108000085 on build GRCh38
Cytoband
4q25
Assay ID Hs04838569_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

3033

Gene Name:

hydroxyacyl-CoA dehydrogenase

Gene Aliases:

HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD

Location:

Chr.4:107989714-108035175 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HADH NM_001184705.2 NP_001171634.2
NM_005327.4 NP_005318.3
AF001902.1 AAB54008.1
AF001903.1 AAB54009.1
AK314444.1
BC000306.2 AAH00306.1
DA825380.1
X96752.1 CAA65528.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1003893 Chr.4:107993422 - 108023690 on Build GRCh38 Gain HADH

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
response to activity
response to insulin
response to drug
negative regulation of insulin secretion
3-hydroxyacyl-CoA dehydrogenase activity
NAD+ binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline