This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04962908_cn
See other CDHR3 CNV Assays ›
Gene Symbol
CDHR3
Assay Reference Genome
Location

Chr.7:105881339 on build GRCh38
Cytoband
7q22.3
Assay ID Hs04962908_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

222256

Gene Name:

cadherin related family member 3

Gene Aliases:

CDH28

Location:

Chr.7:105876773-106036431 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CDHR3 XM_005250224.4 XP_005250281.1
XM_017011863.1 XP_016867352.1
XM_017011865.1 XP_016867354.1
XM_017011866.1 XP_016867355.1
XM_017011867.1 XP_016867356.1
AK125261.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1027213 Chr.7:105798164 - 106382423 on Build GRCh38 Gain NAMPT SYPL1 CDHR3 ATXN7L1
nsv608070 Chr.7:105863755 - 105893289 on Build GRCh38 Loss CDHR3 ATXN7L1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
viral entry into host cell
virus receptor activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline