This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04964208_cn
See other IGF2BP3 CNV Assays ›
Gene Symbol
IGF2BP3
Assay Reference Genome
Location

Chr.7:23331229 on build GRCh38
Cytoband
7p15.3
Assay ID Hs04964208_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10643

Gene Name:

insulin like growth factor 2 mRNA binding protein 3

Gene Aliases:

CT98, IMP-3, IMP3, KOC, KOC1, VICKZ3

Location:

Chr.7:23310209-23470674 on Build GRCh38

Assay Gene Location:

Within Intron 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
IGF2BP3 NM_006547.2 NP_006538.2
XM_006715639.2 XP_006715702.1
XM_011515089.2 XP_011513391.1
XM_011515090.2 XP_011513392.1
XM_011515091.2 XP_011513393.1
XM_011515092.2 XP_011513394.1
XM_011515093.2 XP_011513395.1
AF117108.1 AAD09828.1
BC065269.1 AAH65269.1
BX640800.1 CAE45883.1
BX648488.1 CAH56186.1
U76705.1 AAD09223.1
U97188.1 AAC35208.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv464408 Chr.7:23293751 - 23343516 on Build GRCh38 Loss IGF2BP3 MALSU1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

translation
anatomical structure morphogenesis
gene expression
negative regulation of translation
regulation of cytokine biosynthetic process
mRNA transport
nucleotide binding
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0