This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05061059_cn
See other C8ORF44 CNV Assays ›
Gene Symbol
C8orf44
Assay Reference Genome
Location

Chr.8:66674811 on build GRCh38
Cytoband
8q13.1
Assay ID Hs05061059_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

56260

Gene Name:

chromosome 8 open reading frame 44

Gene Aliases:

-

Location:

Chr.8:66667552-66681149 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
C8orf44 NM_019607.2 NP_062553.1
BC014448.1 AAH14448.1
DA341401.1

Target Gene Details

Entrez Gene ID:

100533105

Gene Name:

C8orf44-SGK3 readthrough

Gene Aliases:

-

Location:

Chr.8:66667552-66862022 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
C8orf44-SGK3 NM_001204173.1 NP_001191102.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831346 Chr.8:66574320 - 66763089 on Build GRCh38 Loss MYBL1 VCPIP1 C8orf44-SGK3 SGK3 C8orf44

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
protein phosphorylation
cellular sodium ion homeostasis
response to stress
positive regulation of sodium ion transport
peptidyl-serine phosphorylation
regulation of cell migration
positive regulation of transporter activity
ion transmembrane transport
intracellular signal transduction
regulation of cell proliferation
regulation of apoptotic process
regulation of sequence-specific DNA binding transcription factor activity
protein kinase activity
protein serine/threonine kinase activity
calcium channel regulator activity
protein binding
ATP binding
potassium channel regulator activity
sodium channel regulator activity
chloride channel regulator activity
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0