This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05107888_cn
See other VTI1A CNV Assays ›
Gene Symbol
VTI1A
Assay Reference Genome
Location

Chr.10:112448726 on build GRCh38
Cytoband
10q25.2
Assay ID Hs05107888_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

143187

Gene Name:

vesicle transport through interaction with t-SNAREs 1A

Gene Aliases:

MMDS3, MVti1, VTI1RP2, Vti1-rp2

Location:

Chr.10:112446988-112947204 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VTI1A NM_001318203.1 NP_001305132.1
NM_001318205.1 NP_001305134.1
NM_145206.3 NP_660207.2
NR_134521.1
NR_134522.1
NR_134523.1
XM_005269544.4 XP_005269601.1
XM_005269545.4 XP_005269602.1
XM_005269546.3 XP_005269603.1
XM_005269547.4 XP_005269604.1
XM_006717637.2 XP_006717700.1
XM_011539328.2 XP_011537630.1
XM_011539330.2 XP_011537632.1
XM_011539331.2 XP_011537633.1
XM_011539333.2 XP_011537635.1
XM_017015745.1 XP_016871234.1
XM_017015746.1 XP_016871235.1
XM_017015747.1 XP_016871236.1
XM_017015748.1 XP_016871237.1
AK095476.1
AK303646.1
BC017052.1 AAH17052.1
BI909044.1
DA324398.1
DA349859.1
DB105695.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831990 Chr.10:112349267 - 112531997 on Build GRCh38 Loss GUCY2GP ZDHHC6 ACSL5 VTI1A
nsv831991 Chr.10:112413098 - 112582139 on Build GRCh38 Loss ZDHHC6 ACSL5 VTI1A

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
autophagy
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
voluntary musculoskeletal movement
Golgi ribbon formation
SNARE binding
SNAP receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-sg6f7:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline