This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05264278_cn
See other FAM118B CNV Assays ›
Gene Symbol
FAM118B
Assay Reference Genome
Location

Chr.11:126233617 on build GRCh38
Cytoband
11q24.2
Assay ID Hs05264278_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79607

Gene Name:

family with sequence similarity 118 member B

Gene Aliases:

-

Location:

Chr.11:126211414-126264540 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FAM118B NM_024556.3 NP_078832.1
XM_005271665.3 XP_005271722.1
XM_011542977.2 XP_011541279.1
XM_011542978.2 XP_011541280.1
XM_017018285.1 XP_016873774.1
XM_017018286.1 XP_016873775.1
AK024756.1 BAB14987.1
AK225081.1
AK315615.1
BC001340.1 AAH01340.1
BC001647.2 AAH01647.1
CB141933.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv556529 Chr.11:126204297 - 126264724 on Build GRCh38 Loss FAM118B RPUSD4 SRPRA
nsv537 Chr.11:126216495 - 126245751 on Build GRCh38 Insertion FAM118B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

Cajal body organization
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline