This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05295651_cn
See other ITGBL1 CNV Assays ›
Gene Symbol
ITGBL1
Assay Reference Genome
Location

Chr.13:101466405 on build GRCh38
Cytoband
13q33.1
Assay ID Hs05295651_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9358

Gene Name:

integrin subunit beta like 1

Gene Aliases:

OSCP, TIED

Location:

Chr.13:101452593-101720857 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ITGBL1 NM_001271754.1 NP_001258683.1
NM_001271755.1 NP_001258684.1
NM_004791.2 NP_004782.1
XM_005254100.4 XP_005254157.1
AB008375.1 BAA22994.1
AF072752.1 AAD17666.1
AK095875.1
AK291340.1
AK297744.1
AK308194.1
BC036788.1 AAH36788.2
BP284376.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv832701 Chr.13:101445422 - 101614006 on Build GRCh38 Loss ITGBL1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

integrin

Gene Ontology Categories:

Function(s)

cell adhesion

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-jmhj8:80/100.66.74.142:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline