This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05419935_cn
See other ABCC11 CNV Assays ›
Gene Symbol
ABCC11
Assay Reference Genome
Location

Chr.16:48172945 on build GRCh38
Cytoband
16q12.1
Assay ID Hs05419935_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

85320

Gene Name:

ATP binding cassette subfamily C member 11

Gene Aliases:

EWWD, MRP8, WW

Location:

Chr.16:48164842-48249973 on Build GRCh38

Assay Gene Location:

Within Intron 33
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ABCC11 NM_032583.3 NP_115972.2
NM_033151.3 NP_149163.2
NM_145186.2 NP_660187.1
XM_011523397.1 XP_011521699.1
XM_011523398.2 XP_011521700.1
XM_017023795.1 XP_016879284.1
XM_017023796.1 XP_016879285.1
XM_017023797.1 XP_016879286.1
XM_017023798.1 XP_016879287.1
XM_017023799.1 XP_016879288.1
XM_017023800.1 XP_016879289.1
XM_017023801.1 XP_016879290.1
XM_017023802.1 XP_016879291.1
AF352582.1 AAK19755.1
AF367202.1 AAK58869.1
AF411579.1 AAL99902.1
AL117406.1
AY040219.1 AAK76739.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

organic anion transport
purine nucleotide transport
transmembrane transport
anion transmembrane transport
ATP binding
organic anion transmembrane transporter activity
purine nucleotide transmembrane transporter activity
anion transmembrane-transporting ATPase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline