This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05476247_cn
See other ALKBH5 CNV Assays ›
Gene Symbol
ALKBH5
Assay Reference Genome
Location

Chr.17:18193879 on build GRCh38
Cytoband
17p11.2
Assay ID Hs05476247_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54890

Gene Name:

alkB homolog 5, RNA demethylase

Gene Aliases:

ABH5, OFOXD, OFOXD1

Location:

Chr.17:18183553-18209954 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ALKBH5 NM_017758.3 NP_060228.3
AK000315.1 BAA91078.1
AK301107.1
AK311161.1
AK311511.1
BC062339.1 AAH62339.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv510702 Chr.17:18134362 - 18213487 on Build GRCh38 Deletion MYO15A ALKBH5

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
positive regulation of defense response to virus by host
mRNA processing
mRNA export from nucleus
spermatogenesis
cell differentiation
oxidative single-stranded RNA demethylation
xenophagy
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
oxidative RNA demethylase activity
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0