This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05479532_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5227009 on build GRCh38
Cytoband
17p13.2
Assay ID Hs05479532_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1

Target Gene Details

Entrez Gene ID:

388325

Gene Name:

SLP adaptor and CSK interacting membrane protein

Gene Aliases:

C17orf87, UNQ5783

Location:

Chr.17:5208920-5234860 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SCIMP NM_001271842.1 NP_001258771.1
NM_001319190.1 NP_001306119.1
NM_207103.3 NP_996986.1
XM_017024615.1 XP_016880104.1
AK057142.1
AK297035.1
AK310145.1
AY358809.1 AAQ89169.1
BC127925.1
BC130554.1
BC130556.1
BC144395.1
DQ778084.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain C17orf107 MIR6865 LOC102724009 INCA1 ARRB2 VMO1 DHX33 CAMTA2 LOC100130950 PSMB6 LOC101559451 LOC400568 LOC107984973 RPAIN LOC101927979 SCIMP ZNF232 GLTPD2 C1QBP CXCL16 PELP1 RABEP1 PLD2 TM4SF5 NUP88 MINK1 SPAG7 GP1BA LOC101928000 USP6 KIF1C ALOX15 ENO3 SLC52A1 MED11 CHRNE ZNF594 ZMYND15 RNF167 SMTNL2 MIR6864 ZFP3 SLC25A11 PFN1
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain ZNF594 SCIMP RABEP1 LOC100130950

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

meiosis I
positive regulation of ERK1 and ERK2 cascade
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0