This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05708319_cn
See other NUP210L CNV Assays ›
Gene Symbol
NUP210L
Assay Reference Genome
Location

Chr.1:154009365 on build GRCh38
Cytoband
1q21.3
Assay ID Hs05708319_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

91181

Gene Name:

nucleoporin 210 like

Gene Aliases:

-

Location:

Chr.1:153992582-154155116 on Build GRCh38

Assay Gene Location:

Within Intron 35
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NUP210L NM_001159484.1 NP_001152956.1
NM_207308.2 NP_997191.2
XM_011510122.1 XP_011508424.1
XM_011510123.1 XP_011508425.1
XM_011510124.1 XP_011508426.1
XM_017002788.1 XP_016858277.1
XM_017002789.1 XP_016858278.1
AK125924.1 BAC86345.1
AK128108.1 BAC87279.1
AL133633.1 CAB63757.1
AL833964.1 CAD38812.1
BX648132.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s)

spermatid development
Sertoli cell development

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline