This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05726532_cn
See other ADAM15 CNV Assays ›
Gene Symbol
ADAM15
Assay Reference Genome
Location

Chr.1:155051470 on build GRCh38
Cytoband
1q21.3
Assay ID Hs05726532_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8751

Gene Name:

ADAM metallopeptidase domain 15

Gene Aliases:

MDC15

Location:

Chr.1:155051272-155062776 on Build GRCh38

Assay Gene Location:

Overlaps Exon 1 - Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ADAM15 NM_001261464.1 NP_001248393.1
NM_001261465.1 NP_001248394.1
NM_001261466.1 NP_001248395.1
NM_003815.4 NP_003806.3
NM_207191.2 NP_997074.1
NM_207194.2 NP_997077.1
NM_207195.2 NP_997078.1
NM_207196.2 NP_997079.1
NM_207197.2 NP_997080.1
NR_048577.1
NR_048578.1
NR_048579.1
AB209157.1 1 525 BAD92394.1
AK000667.1
AK075498.1
AK295614.1
AK296925.1
AK297468.1
AK308172.1
AY518542.1 AAR99331.1
AY560593.1 AAS72991.1
AY560594.1 AAS72992.1
AY560595.1 AAS72993.1
AY560596.1 AAS72994.1
AY560597.1 AAS72995.1
AY560598.1 AAS72996.1
AY560599.1 AAS72997.1
AY560600.1 AAS72998.1
AY560601.1 AAS72999.1
AY576417.1 AAS73000.1
BC014566.2 AAH14566.1
BT009764.1 AAP88766.1
U41767.1 AAC50404.1
U46005.1 AAC51112.1

Target Gene Details

Entrez Gene ID:

100505666

Gene Name:

uncharacterized LOC100505666

Gene Aliases:

-

Location:

Chr.1:155045191-155063991 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100505666 NR_040773.1
BM047608.1
DB152307.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv509502 Chr.1:154904407 - 155142291 on Build GRCh38 Insertion PBXIP1 EFNA4 EFNA3 ZBTB7B DPM3 PMVK LOC100505666 FLAD1 ADAM15 LENEP SHC1 LOC101928120 MIR4258 SLC50A1 DCST2 CKS1B EFNA1 PYGO2 DCST1
esv3587615 Chr.1:155047853 - 155175013 on Build GRCh38 Gain EFNA4 EFNA3 DPM3 LOC100505666 EFNA1 TRIM46 ADAM15 KRTCAP2 DCST1 SLC50A1
nsv952212 Chr.1:154998225 - 155087624 on Build GRCh38 Deletion EFNA4 EFNA3 ZBTB7B LOC100505666 ADAM15 DCST1 DCST2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
negative regulation of cell-matrix adhesion
proteolysis
apoptotic process
cell-matrix adhesion
integrin-mediated signaling pathway
male gonad development
extracellular matrix disassembly
negative regulation of cell growth
negative regulation of cell migration
collagen catabolic process
tissue regeneration
innate immune response
cardiac epithelial to mesenchymal transition
protein kinase C signaling
negative regulation of receptor binding
metalloendopeptidase activity
integrin binding
protein binding
metallopeptidase activity
zinc ion binding
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-b64d564dd-lbfdq:80/100.66.77.223:80.
git-commit: 96463e515cbe20f5bdbdc562e005fc2cf160896a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.2-2025.08.98-1.0