This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05745980_cn
See other HMGCL CNV Assays ›
Gene Symbol
HMGCL
Assay Reference Genome
Location

Chr.1:23824141 on build GRCh38
Cytoband
1p36.11
Assay ID Hs05745980_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

3155

Gene Name:

3-hydroxymethyl-3-methylglutaryl-CoA lyase

Gene Aliases:

HL

Location:

Chr.1:23801877-23825459 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HMGCL NM_000191.2 NP_000182.2
NM_001166059.1 NP_001159531.1
AK122801.1
AK300733.1
AK309129.1
AK313869.1
BC010570.1 AAH10570.1
BT009792.1 AAP88794.1
CR456884.1 CAG33165.1
DA425822.1
L07033.1 AAA92733.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

liver development
leucine catabolic process
acyl-CoA metabolic process
mitochondrion organization
response to nutrient
response to starvation
ketone body biosynthetic process
protein tetramerization
response to fatty acid
fatty-acyl-CoA binding
magnesium ion binding
hydroxymethylglutaryl-CoA lyase activity
receptor binding
manganese ion binding
carboxylic acid binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0