This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05748156_cn
See other FGGY CNV Assays ›
Gene Symbol
FGGY
Assay Reference Genome
Location

Chr.1:59314398 on build GRCh38
Cytoband
1p32.1
Assay ID Hs05748156_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

55277

Gene Name:

FGGY carbohydrate kinase domain containing

Gene Aliases:

-

Location:

Chr.1:59296405-59762730 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FGGY NM_001113411.1 NP_001106882.1
NM_001244714.1 NP_001231643.1
NM_018291.3 NP_060761.3
NR_103473.1
XM_011541730.1 XP_011540032.1
XM_011541731.1 XP_011540033.1
XM_011541732.1 XP_011540034.1
XM_011541736.2 XP_011540038.1
XM_017001643.1 XP_016857132.1
XM_017001644.1 XP_016857133.1
XM_017001645.1 XP_016857134.1
XM_017001646.1 XP_016857135.1
XM_017001647.1 XP_016857136.1
XM_017001649.1 XP_016857138.1
XM_017001650.1 XP_016857139.1
XM_017001651.1 XP_016857140.1
XM_017001652.1 XP_016857141.1
XM_017001653.1 XP_016857142.1
XM_017001654.1 XP_016857143.1
XM_017001655.1 XP_016857144.1
XM_017001656.1 XP_016857145.1
XM_017001659.1 XP_016857148.1
XM_017001660.1 XP_016857149.1
XM_017001661.1 XP_016857150.1
XM_017001663.1 XP_016857152.1
XM_017001664.1 XP_016857153.1
XM_017001665.1 XP_016857154.1
XM_017001666.1 XP_016857155.1
XM_017001667.1 XP_016857156.1
XM_017001669.1 XP_016857158.1
XM_017001671.1 XP_016857160.1
XM_017001672.1 XP_016857161.1
XM_017001673.1 XP_016857162.1
XM_017001674.1 XP_016857163.1
XM_017001675.1 XP_016857164.1
XM_017001676.1 XP_016857165.1
XM_017001679.1 XP_016857168.1
AK022237.1 BAB13993.1
AK054842.1
AK090568.1
AK315649.1
BC014947.1 AAH14947.1
BX647626.1
DA072156.1
DA969344.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv829982 Chr.1:59247176 - 59367120 on Build GRCh38 Loss FGGY

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

carbohydrate kinase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0