This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05761635_cn
See other SPEN CNV Assays ›
Gene Symbol
SPEN
Assay Reference Genome
Location

Chr.1:15883060 on build GRCh38
Cytoband
1p36.21
Assay ID Hs05761635_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

23013

Gene Name:

spen family transcriptional repressor

Gene Aliases:

HIAA0929, MINT, RBM15C, SHARP

Location:

Chr.1:15847864-15940455 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SPEN NM_015001.2 NP_055816.2
AF356524.1 AAK52750.1
AK127577.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833647 Chr.1:15794042 - 15952334 on Build GRCh38 Loss UQCRHL SPEN FLJ37453 ZBTB17
dgv37n100 Chr.1:15823408 - 15924038 on Build GRCh38 Gain SPEN FLJ37453
dgv38n100 Chr.1:15839166 - 15924038 on Build GRCh38 Gain SPEN FLJ37453

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
mRNA splicing, via spliceosome
transcription, DNA-templated
Notch signaling pathway
viral process
negative regulation of transcription, DNA-templated
positive regulation of neurogenesis
nucleotide binding
RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
nucleic acid binding
DNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0