This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05769432_cn
See other UCHL5 CNV Assays ›
Gene Symbol
UCHL5
Assay Reference Genome
Location

Chr.1:193051097 on build GRCh38
Cytoband
1q31.2
Assay ID Hs05769432_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51377

Gene Name:

ubiquitin C-terminal hydrolase L5

Gene Aliases:

CGI-70, INO80R, UCH-L5, UCH37

Location:

Chr.1:193012366-193060140 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UCHL5 NM_001199261.1 NP_001186190.1
NM_001199262.1 NP_001186191.1
NM_001199263.1 NP_001186192.1
NM_015984.3 NP_057068.1
NR_037607.1
XM_005245244.4 XP_005245301.1
XM_005245246.4 XP_005245303.1
XM_005245247.4 XP_005245304.1
XM_005245248.4 XP_005245305.1
XM_005245249.4 XP_005245306.1
XM_006711366.3 XP_006711429.1
XM_006711367.3 XP_006711430.1
XM_006711368.3 XP_006711431.1
XM_006711369.2 XP_006711432.1
XM_006711370.3 XP_006711433.3
XM_006711371.3 XP_006711434.1
XM_011509604.1 XP_011507906.1
XM_011509607.2 XP_011507909.1
XM_011509608.2 XP_011507910.1
XM_011509609.2 XP_011507911.1
XM_017001429.1 XP_016856918.1
XM_017001430.1 XP_016856919.1
XM_017001431.1 XP_016856920.1
XM_017001432.1 XP_016856921.1
XM_017001433.1 XP_016856922.1
XM_017001434.1 XP_016856923.1
XM_017001435.1 XP_016856924.1
XM_017001436.1 XP_016856925.1
XM_017001437.1 XP_016856926.1
XM_017001438.1 XP_016856927.1
XM_017001439.1 XP_016856928.1
XM_017001440.1 XP_016856929.1
XM_017001441.1 XP_016856930.1
XM_017001442.1 XP_016856931.1
XM_017001443.1 XP_016856932.1
XM_017001444.1 XP_016856933.1
AF147717.1 AAD31528.1
AF151828.1 AAD34065.1
AF157320.1 AAF67486.1
AK024751.1
AK225794.1
AK296487.1
AK301381.1
AK314445.1
AK316064.1
BC015521.1 AAH15521.1
BC025369.2 AAH25369.1
BT006790.1 AAP35436.1
DA255130.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv498n100 Chr.1:192992568 - 193114915 on Build GRCh38 Loss SCARNA18B TROVE2 UCHL5 GLRX2
nsv466994 Chr.1:192394185 - 193552205 on Build GRCh38 Loss MIR1278 RGS13 TROVE2 B3GALT2 LINC01032 CDC73 SCARNA18B RGS2 UCHL5 LINC01031 GLRX2 RGS1 MIR4426

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
thiol-dependent ubiquitin-specific protease activity
endopeptidase inhibitor activity
protein binding
poly(A) RNA binding
proteasome binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0