This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05772935_cn
See other PRDM16 CNV Assays ›
Gene Symbol
PRDM16
Assay Reference Genome
Location

Chr.1:3070812 on build GRCh38
Cytoband
1p36.32
Assay ID Hs05772935_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

63976

Gene Name:

PR domain 16

Gene Aliases:

CMD1LL, LVNC8, MEL1, PFM13

Location:

Chr.1:3069025-3438621 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PRDM16 NM_022114.3 NP_071397.3
NM_199454.2 NP_955533.2
XM_005244772.4 XP_005244829.1
XM_005244773.4 XP_005244830.1
XM_005244774.4 XP_005244831.1
XM_006710814.3 XP_006710877.1
XM_017002050.1 XP_016857539.1
AB051462.2
AB078876.1 BAB84297.1
AF294278.1 AAG33382.1
BC161614.1
BQ897896.1
BX092842.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3585036 Chr.1:3004067 - 3089193 on Build GRCh38 Gain PRDM16 ACTRT2 LINC00982
nsv829759 Chr.1:3006585 - 3094573 on Build GRCh38 Loss PRDM16 ACTRT2 LINC00982
nsv824697 Chr.1:3059293 - 3078148 on Build GRCh38 Gain PRDM16 LINC00982
nsv951959 Chr.1:3051037 - 3812036 on Build GRCh38 Deletion TP73 PRDM16 LRRC47 TPRG1L TP73-AS1 LINC00982 MIR4251 WRAP73 SMIM1 CCDC27 MIR551A LOC105378604 ARHGEF16 MEGF6
nsv545098 Chr.1:3009815 - 3089567 on Build GRCh38 Loss PRDM16 ACTRT2 LINC00982
nsv1013792 Chr.1:3005296 - 3391724 on Build GRCh38 Gain MIR4251 PRDM16 ACTRT2 LOC105378604 LINC00982
nsv1000306 Chr.1:3031140 - 3164376 on Build GRCh38 Gain MIR4251 PRDM16 LINC00982
esv3583359 Chr.1:2777970 - 3286320 on Build GRCh38 Gain MIR4251 PRDM16 TTC34 ACTRT2 LINC00982
nsv1076898 Chr.1:3069235 - 3070836 on Build GRCh38 Deletion PRDM16
dgv11n100 Chr.1:2777886 - 3350773 on Build GRCh38 Gain MIR4251 PRDM16 TTC34 ACTRT2 LOC105378604 LINC00982
nsv428399 Chr.1:2622539 - 3235926 on Build GRCh38 Gain MIR4251 PRDM16 TTC34 MMEL1 ACTRT2 LINC00982
nsv1142739 Chr.1:3070136 - 3070836 on Build GRCh38 Deletion PRDM16

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
neurogenesis
negative regulation of transforming growth factor beta receptor signaling pathway
histone lysine methylation
somatic stem cell population maintenance
regulation of cellular respiration
tongue development
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
white fat cell differentiation
brown fat cell differentiation
palate development
positive regulation of brown fat cell differentiation
transcription coactivator activity
protein binding
histone-lysine N-methyltransferase activity
activating transcription factor binding
sequence-specific DNA binding
SMAD binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-5c659bcd8-c4h4v:80/100.66.79.7:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline