This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05786103_cn
See other TIE1 CNV Assays ›
Gene Symbol
TIE1
Assay Reference Genome
Location

Chr.1:43304301 on build GRCh38
Cytoband
1p34.2
Assay ID Hs05786103_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

7075

Gene Name:

tyrosine kinase with immunoglobulin like and EGF like domains 1

Gene Aliases:

JTK14, TIE

Location:

Chr.1:43300895-43323110 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
TIE1 NM_001253357.1 NP_001240286.1
NM_005424.4 NP_005415.1
XM_005271163.1 XP_005271220.1
XM_006710869.1 XP_006710932.1
XM_017002207.1 XP_016857696.1
AF217976.1 AAG17219.1
AK225560.1
AK300399.1
AK300623.1
AK307639.1
BC038239.1 AAH38239.1
DB287371.1
EU826586.1
EU826587.1
EU826588.1
EU826589.1
EU826590.1
X60957.1 CAA43290.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv546128 Chr.1:43301467 - 43451738 on Build GRCh38 Loss MED8 SZT2 HYI MIR6735 MPL CDC20 TIE1 ELOVL1 MIR6734
nsv829693 Chr.1:43129457 - 43312977 on Build GRCh38 Loss FAM183A EBNA1BP2 TMEM125 C1orf210 TIE1 MIR6733 CFAP57

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
vasculogenesis
in utero embryonic development
signal transduction
mesoderm development
negative regulation of angiogenesis
peptidyl-tyrosine phosphorylation
negative regulation of cell migration
response to retinoic acid
plasma membrane fusion
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline