This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05787016_cn
See other PRPF3 CNV Assays ›
Gene Symbol
PRPF3
Assay Reference Genome
Location

Chr.1:150321929 on build GRCh38
Cytoband
1q21.2
Assay ID Hs05787016_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9129

Gene Name:

pre-mRNA processing factor 3

Gene Aliases:

HPRP3, HPRP3P, PRP3, Prp3p, RP18, SNRNP90

Location:

Chr.1:150321479-150353233 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PRPF3 NM_004698.2 NP_004689.1
XM_011510129.1 XP_011508431.1
XM_011510131.2 XP_011508433.1
XM_011510132.2 XP_011508434.1
XM_017002791.1 XP_016858280.1
AF016370.1 AAC51926.1
AK222825.1 BAD96545.1
AK297139.1
AK297265.1
AK299980.1
AK309330.1
AK310241.1
AK311653.1
AK312891.1
DC407158.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831548 Chr.1:150296285 - 150463240 on Build GRCh38 Loss RPRD2 MRPS21 PRPF3

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal tri-snRNP complex assembly
RNA splicing, via transesterification reactions
mRNA splicing, via spliceosome
mRNA processing
RNA splicing
protein binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0