This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05799316_cn
See other CACHD1 CNV Assays ›
Gene Symbol
CACHD1
Assay Reference Genome
Location

Chr.1:64498268 on build GRCh38
Cytoband
1p31.3
Assay ID Hs05799316_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

57685

Gene Name:

cache domain containing 1

Gene Aliases:

-

Location:

Chr.1:64470793-64693058 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CACHD1 NM_020925.2 NP_065976.2
CX163220.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv546437 Chr.1:64230394 - 64538095 on Build GRCh38 Loss CACHD1 UBE2U
esv2660804 Chr.1:64381364 - 64626908 on Build GRCh38 Deletion MIR4794 CACHD1
nsv1000188 Chr.1:64263100 - 64546875 on Build GRCh38 Gain CACHD1 UBE2U
nsv830048 Chr.1:64367523 - 64545238 on Build GRCh38 Gain CACHD1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s)

calcium ion transport

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline