This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05802817_cn
See other GTF2B CNV Assays ›
Gene Symbol
GTF2B
Assay Reference Genome
Location

Chr.1:88874120 on build GRCh38
Cytoband
1p22.2
Assay ID Hs05802817_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2959

Gene Name:

general transcription factor IIB

Gene Aliases:

TF2B, TFIIB

Location:

Chr.1:88852638-88891944 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GTF2B NM_001514.5 NP_001505.1
XM_011541299.1 XP_011539601.1
AB451296.1
AK289822.1
BC020597.1 AAH20597.1
BP379342.1
F07886.1
M76766.1 AAA61149.1
S44184.1 AAB23144.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830537 Chr.1:88790384 - 88940395 on Build GRCh38 Gain GTF2B PKN2 KYAT3
nsv830525 Chr.1:88776863 - 88975647 on Build GRCh38 Loss GTF2B PKN2 KYAT3
nsv830514 Chr.1:88715143 - 88876830 on Build GRCh38 Loss GTF2B PKN2
dgv22n27 Chr.1:88791695 - 88894837 on Build GRCh38 Gain GTF2B PKN2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
viral process
snRNA transcription from RNA polymerase II promoter
positive regulation of viral transcription
core promoter binding
protein binding
transcription factor binding
zinc ion binding
TBP-class protein binding
thyroid hormone receptor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0