This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05805641_cn
See other PATJ CNV Assays ›
Gene Symbol
PATJ
Assay Reference Genome
Location

Chr.1:61750806 on build GRCh38
Cytoband
1p31.3
Assay ID Hs05805641_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10207

Gene Name:

PATJ, crumbs cell polarity complex component

Gene Aliases:

Cipp, INADL, InaD-like, hINADL

Location:

Chr.1:61742477-62163920 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PATJ NM_176877.2 NP_795352.2
XM_005270341.3 XP_005270398.1
XM_005270347.1 XP_005270404.1
XM_006710278.3 XP_006710341.1
XM_011540462.2 XP_011538764.1
XM_011540463.2 XP_011538765.1
XM_011540464.2 XP_011538766.1
XM_011540465.2 XP_011538767.1
XM_011540466.2 XP_011538768.1
XM_011540467.2 XP_011538769.1
XM_011540468.2 XP_011538770.1
XM_011540469.2 XP_011538771.1
XM_011540470.1 XP_011538772.1
XM_016999998.1 XP_016855487.1
XM_016999999.1 XP_016855488.1
AJ001306.1 CAA04666.1
AJ224747.1 CAA12112.1
AJ224748.1 CAA12113.1
AK307713.1
AK308428.1
BC142661.1
BC142723.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

intracellular signal transduction
bicellular tight junction assembly
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-pcbtx:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0