This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05806134_cn
See other PRDM2 CNV Assays ›
Gene Symbol
PRDM2
Assay Reference Genome
Location

Chr.1:13733967 on build GRCh38
Cytoband
1p36.21
Assay ID Hs05806134_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

7799

Gene Name:

PR domain 2

Gene Aliases:

HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2

Location:

Chr.1:13698875-13825079 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PRDM2 NM_001135610.1 NP_001129082.1
NM_012231.4 NP_036363.2
NM_015866.4 NP_056950.2
XM_017002253.1 XP_016857742.1
XM_017002254.1 XP_016857743.1
XM_017002255.1 XP_016857744.1
XM_017002256.1 XP_016857745.1
XM_017002257.1 XP_016857746.1
XM_017002258.1 XP_016857747.1
XM_017002264.1 XP_016857753.1
U17838.1 AAC50820.2

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv545567 Chr.1:13558784 - 14026576 on Build GRCh38 Gain PRDM2 PDPN KAZN

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
determination of adult lifespan
response to estradiol
histone lysine methylation
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
histone-lysine N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-97vht:80/100.66.77.199:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline