This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05876700_cn
See other RPE CNV Assays ›
Gene Symbol
RPE
Assay Reference Genome
Location

Chr.2:210005218 on build GRCh38
Cytoband
2q34
Assay ID Hs05876700_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

6120

Gene Name:

ribulose-5-phosphate-3-epimerase

Gene Aliases:

RPE2-1

Location:

Chr.2:210002565-210029156 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RPE NM_001278282.1 NP_001265211.1
NM_001278283.1 NP_001265212.1
NM_001278285.1 NP_001265214.1
NM_001278286.1 NP_001265215.1
NM_001278288.1 NP_001265217.1
NM_001278289.1 NP_001265218.1
NM_001318926.1 NP_001305855.1
NM_001318927.1 NP_001305856.1
NM_001318928.1 NP_001305857.1
NM_001318929.1 NP_001305858.1
NM_001318930.1 NP_001305859.1
NM_001318931.1 NP_001305860.1
NM_006916.2 NP_008847.1
NM_199229.2 NP_954699.1
XM_006712677.3 XP_006712740.1
AK056028.1 BAB71076.1
AK093658.1 BAC04212.1
AK096199.1
AK291035.1
AK303184.1
AK308741.1
AL833120.1
BC005148.2 AAH05148.2
BC016764.1 AAH16764.1
BC072401.1 AAH72401.1
BG546879.1
BX403091.2
BX643719.1
CN372165.1
CR992340.1
DA237285.1
DA336224.1
DA592494.1
DC403549.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
pentose-phosphate shunt
pentose-phosphate shunt, non-oxidative branch
pentose catabolic process
cellular carbohydrate metabolic process
ribulose-phosphate 3-epimerase activity
identical protein binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0