This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05900776_cn
See other P3H2-AS1 CNV Assays ›
Gene Symbol
P3H2-AS1
Assay Reference Genome
Location

Chr.3:190130249 on build GRCh38
Cytoband
3q28
Assay ID Hs05900776_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

101929152

Gene Name:

P3H2 antisense RNA 1

Gene Aliases:

LEPREL1-AS1

Location:

Chr.3:190120964-190144846 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
P3H2-AS1 NR_126419.1
BM017065.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1014822 Chr.3:190065059 - 190464556 on Build GRCh38 Gain CLDN16 CLDN1 TMEM207 P3H2-AS1 P3H2
esv3893843 Chr.3:190075998 - 191094501 on Build GRCh38 Gain SNAR-I CLDN16 CLDN1 IL1RAP GMNC TMEM207 P3H2-AS1 P3H2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0