This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05990969_cn
See other C5ORF66 CNV Assays ›
Gene Symbol
C5orf66
Assay Reference Genome
Location

Chr.5:135041123 on build GRCh38
Cytoband
5q31.1
Assay ID Hs05990969_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100996485

Gene Name:

chromosome 5 open reading frame 66

Gene Aliases:

-

Location:

Chr.5:135033280-135344680 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
C5orf66 NM_001277348.1 NP_001264277.1
AK026965.1 BAB15606.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv823239 Chr.5:135019535 - 135071110 on Build GRCh38 Gain C5orf66-AS1 C5orf66 PITX1
nsv830495 Chr.5:135024595 - 135204207 on Build GRCh38 Loss C5orf66-AS1 C5orf66 PITX1
nsv823240 Chr.5:135020631 - 135043367 on Build GRCh38 Gain C5orf66-AS1 C5orf66 PITX1
dgv969n67 Chr.5:135018061 - 135059172 on Build GRCh38 Gain C5orf66-AS1 C5orf66 PITX1
nsv599744 Chr.5:135012266 - 135070292 on Build GRCh38 Loss C5orf66-AS1 C5orf66 PITX1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4sh5g:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline