This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06059058_cn
See other CAMK4 CNV Assays ›
Gene Symbol
CAMK4
Assay Reference Genome
Location

Chr.5:111237722 on build GRCh38
Cytoband
5q22.1
Assay ID Hs06059058_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

814

Gene Name:

calcium/calmodulin dependent protein kinase IV

Gene Aliases:

CaMK IV, CaMK-GR, caMK

Location:

Chr.5:111223653-111498503 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CAMK4 NM_001323374.1 NP_001310303.1
NM_001323375.1 NP_001310304.1
NM_001323376.1 NP_001310305.1
NM_001323377.1 NP_001310306.1
NM_001744.5 NP_001735.1
AK311662.1
AK312840.1
BC016695.1 AAH16695.2
BC025687.1 AAH25687.1
BG721313.1
D30742.1 BAA06403.1
DA114495.1
DA396146.1
L17000.1 AAA35639.1
L24959.1 AAA18251.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3606259 Chr.5:110985051 - 111262481 on Build GRCh38 Loss TSLP CAMK4 WDR36

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
myeloid dendritic cell cytokine production
protein phosphorylation
inflammatory response
signal transduction
long-term memory
peptidyl-serine phosphorylation
regulation of T cell differentiation in thymus
intracellular signal transduction
myeloid dendritic cell differentiation
regulation of osteoclast differentiation
positive regulation of transcription, DNA-templated
protein autophosphorylation
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline