This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06275987_cn
See other CPSF7 CNV Assays ›
Gene Symbol
CPSF7
Assay Reference Genome
Location

Chr.11:61413839 on build GRCh38
Cytoband
11q12.2
Assay ID Hs06275987_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79869

Gene Name:

cleavage and polyadenylation specific factor 7

Gene Aliases:

CFIm59

Location:

Chr.11:61402648-61429992 on Build GRCh38

Assay Gene Location:

Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF7 NM_001136040.2 NP_001129512.1
NM_001142565.1 NP_001136037.1
NM_024811.3 NP_079087.3
XM_005274298.4 XP_005274355.1
XM_005274299.4 XP_005274356.1
XM_005274303.4 XP_005274360.1
XM_011545257.2 XP_011543559.1
XM_011545258.2 XP_011543560.1
XM_011545259.2 XP_011543561.1
XM_011545260.2 XP_011543562.1
XM_011545261.2 XP_011543563.1
XM_011545262.2 XP_011543564.1
XM_011545263.2 XP_011543565.1
XM_017018345.1 XP_016873834.1
AJ275970.1 CAC81661.1
AK022591.1 BAB14118.1
AK092037.1
AK096343.1
AK294578.1
AL512759.1 CAC21678.1
AL832546.1
AL832667.1
AL833332.1
BC018135.1 AAH18135.1
BX537888.1 CAD97884.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv520751 Chr.11:61389584 - 61415916 on Build GRCh38 Gain CPSF7 TMEM216
nsv832178 Chr.11:61402074 - 61573390 on Build GRCh38 Gain+Loss MIR4488 SYT7 SDHAF2 PPP1R32 CPSF7 LRRC10B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA 3'-end processing
protein tetramerization
nucleotide binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline