This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06292765_cn
See other MS4A1 CNV Assays ›
Gene Symbol
MS4A1
Assay Reference Genome
Location

Chr.11:60459862 on build GRCh38
Cytoband
11q12.2
Assay ID Hs06292765_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

931

Gene Name:

membrane spanning 4-domains A1

Gene Aliases:

B1, Bp35, CD20, CVID5, LEU-16, MS4A2, S7

Location:

Chr.11:60455809-60470752 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MS4A1 NM_021950.3 NP_068769.2
NM_152866.2 NP_690605.1
AK225630.1
AK292168.1
AK300025.1
AK310118.1
AW504475.1
BC002807.2 AAH02807.1
CD691924.1
X07203.1 CAA30179.1
X12530.1 CAA31046.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2744569 Chr.11:60310239 - 60485227 on Build GRCh38 Deletion MS4A5 MS4A1 MS4A7 MS4A6E MS4A14
dgv51n111 Chr.11:60447593 - 60463593 on Build GRCh38 Deletion MS4A5 MS4A1
nsv508635 Chr.11:60422385 - 60487633 on Build GRCh38 Deletion MS4A5 MS4A1
nsv555151 Chr.11:60448972 - 60472250 on Build GRCh38 Loss MS4A1
dgv391e212 Chr.11:60455519 - 60463836 on Build GRCh38 Loss MS4A1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

humoral immune response
B cell proliferation
epidermal growth factor receptor binding
protein binding
MHC class II protein complex binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline