This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06293962_cn
See other UBASH3B CNV Assays ›
Gene Symbol
UBASH3B
Assay Reference Genome
Location

Chr.11:122670650 on build GRCh38
Cytoband
11q24.1
Assay ID Hs06293962_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

84959

Gene Name:

ubiquitin associated and SH3 domain containing B

Gene Aliases:

STS-1, STS1, TULA-2, TULA2, p70

Location:

Chr.11:122655690-122814479 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBASH3B NM_032873.4 NP_116262.2
XM_011543042.1 XP_011541344.1
AB075839.1
AK075203.1 BAC11468.1
AK222843.1 BAD96563.1
AK222846.1 BAD96566.1
BC007541.1 AAH07541.2
DA197468.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1053418 Chr.11:122662739 - 122818935 on Build GRCh38 Gain UBASH3B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
negative regulation of signal transduction
peptidyl-tyrosine dephosphorylation
negative regulation of osteoclast differentiation
negative regulation of bone resorption
regulation of release of sequestered calcium ion into cytosol
negative regulation of platelet aggregation
protein tyrosine phosphatase activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline