This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06340763_cn
See other CACNA1C CNV Assays ›
Gene Symbol
CACNA1C
Assay Reference Genome
Location

Chr.12:1987185 on build GRCh38
Cytoband
12p13.33
Assay ID Hs06340763_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

775

Gene Name:

calcium voltage-gated channel subunit alpha1 C

Gene Aliases:

CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS

Location:

Chr.12:1969677-2697949 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CACNA1C XM_006719017.2 XP_006719080.1
XM_011521020.2 XP_011519322.1
XM_017019926.1 XP_016875415.1
XM_017019927.1 XP_016875416.1
XM_017019928.1 XP_016875417.1
XM_017019929.1 XP_016875418.1
XM_017019930.1 XP_016875419.1
XM_017019931.1 XP_016875420.1
XM_017019932.1 XP_016875421.1
XM_017019933.1 XP_016875422.1
XM_017019934.1 XP_016875423.1
XM_017019935.1 XP_016875424.1
XM_017019936.1 XP_016875425.1
XM_017019937.1 XP_016875426.1
XM_017019938.1 XP_016875427.1
XM_017019939.1 XP_016875428.1
XM_017019940.1 XP_016875429.1
XM_017019941.1 XP_016875430.1
XM_017019942.1 XP_016875431.1
XM_017019943.1 XP_016875432.1
XM_017019944.1 XP_016875433.1
XM_017019945.1 XP_016875434.1
XM_017019946.1 XP_016875435.1
XM_017019947.1 XP_016875436.1
XM_017019948.1 XP_016875437.1
XM_017019949.1 XP_016875438.1
XM_017019950.1 XP_016875439.1
XM_017019951.1 XP_016875440.1
XM_017019952.1 XP_016875441.1
XM_017019953.1 XP_016875442.1
XM_017019954.1 XP_016875443.1
XM_017019955.1 XP_016875444.1

Target Gene Details

Entrez Gene ID:

196513

Gene Name:

decapping mRNA 1B

Gene Aliases:

DCP1

Location:

Chr.12:1941591-2004535 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DCP1B NM_152640.4 NP_689853.3
NR_135060.1
XM_011520927.2 XP_011519229.1
AK056200.1 BAB71118.1
AK299203.1
AK310475.1
AY146652.1 AAN62764.1
BC043437.2 AAH43437.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv521838 Chr.12:1951414 - 2190514 on Build GRCh38 Gain DCP1B CACNA1C CACNA1C-IT2
nsv1038533 Chr.12:1974559 - 2000280 on Build GRCh38 Gain DCP1B CACNA1C
nsv1042378 Chr.12:1968731 - 2020607 on Build GRCh38 Gain DCP1B CACNA1C
nsv1040370 Chr.12:1912229 - 2561851 on Build GRCh38 Gain DCP1B LINC00940 CACNA1C CACNA1C-IT3 CACNA2D4 CACNA1C-AS4 CACNA1C-IT2
nsv525455 Chr.12:1956204 - 2025860 on Build GRCh38 Loss DCP1B CACNA1C

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

voltage-gated ion channel mRNA capping factor

Gene Ontology Categories:

Function(s) Process(es)

immune system development
positive regulation of cytosolic calcium ion concentration
heart development
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
embryonic forelimb morphogenesis
calcium-mediated signaling using extracellular calcium source
camera-type eye development
regulation of insulin secretion
calcium ion transport into cytosol
cardiac conduction
calcium ion transmembrane transport via high voltage-gated calcium channel
calcium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during AV node cell action potential
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
membrane depolarization during atrial cardiac muscle cell action potential
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
deadenylation-dependent decapping of nuclear-transcribed mRNA
deadenylation-independent decapping of nuclear-transcribed mRNA
positive regulation of catalytic activity
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
voltage-gated calcium channel activity
protein binding
calmodulin binding
high voltage-gated calcium channel activity
metal ion binding
alpha-actinin binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
mRNA binding
enzyme activator activity
hydrolase activity
enzyme regulator activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0