This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06449898_cn
See other AQP4-AS1 CNV Assays ›
Gene Symbol
AQP4-AS1
Assay Reference Genome
Location

Chr.18:26894526 on build GRCh38
Cytoband
18q11.2
Assay ID Hs06449898_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

147429

Gene Name:

AQP4 antisense RNA 1

Gene Aliases:

C18orf16, CHST9-AS1

Location:

Chr.18:26865308-26935946 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AQP4-AS1 NR_026908.1
AK055069.1 BAB70850.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2761995 Chr.18:26634322 - 26922606 on Build GRCh38 Loss KCTD1 AQP4-AS1 AQP4 CHST9 PCAT18
nsv833600 Chr.18:26893930 - 27101055 on Build GRCh38 Gain AQP4-AS1 CHST9

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline