This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06508801_cn
See other NDUFS5 CNV Assays ›
Gene Symbol
NDUFS5
Assay Reference Genome
Location

Chr.1:39033112 on build GRCh38
Cytoband
1p34.3
Assay ID Hs06508801_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4725

Gene Name:

NADH:ubiquinone oxidoreductase subunit S5

Gene Aliases:

CI-15k, CI15K

Location:

Chr.1:39026295-39034636 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NDUFS5 NM_001184979.1 NP_001171908.1
NM_004552.2 NP_004543.1
AF020352.1 AAB87866.1
AF047434.1 AAC39878.1
AF086268.1
AI244607.1
AK312188.1
BC001884.1 AAH01884.1
BC093057.1 AAH93057.1
CR456904.1 CAG33185.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3585778 Chr.1:39021596 - 39033061 on Build GRCh38 Gain NDUFS5

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0