This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06513232_cn
See other NFYC CNV Assays ›
Gene Symbol
NFYC
Assay Reference Genome
Location

Chr.1:40713337 on build GRCh38
Cytoband
1p34.2
Assay ID Hs06513232_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4802

Gene Name:

nuclear transcription factor Y subunit gamma

Gene Aliases:

CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC

Location:

Chr.1:40691570-40771603 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NFYC NM_001142587.1 NP_001136059.1
NM_001142588.1 NP_001136060.1
NM_001142589.1 NP_001136061.1
NM_001142590.1 NP_001136062.1
NM_001308115.1 NP_001295044.1
NM_014223.4 NP_055038.2
XM_005270894.1 XP_005270951.1
XM_006710661.1 XP_006710724.1
XM_017001365.1 XP_016856854.1
XM_017001367.1 XP_016856856.1
XM_017001368.1 XP_016856857.1
AB208975.1 BAD92212.1
AK000346.1 BAA91100.1
AK127677.1
AK300774.1
AK301215.1
AK301385.1
BC005003.1 AAH05003.1
BP230898.1
D85425.1 BAA12818.1
D89986.1 BAA14051.1
DC297902.1
DC391720.1
DC428152.1
U78774.1 AAC51669.1
Z70024.1 CAA93846.1
Z74792.1 CAA99055.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein folding
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0