This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06513829_cn
See other ABCA4 CNV Assays ›
Gene Symbol
ABCA4
Assay Reference Genome
Location

Chr.1:94012777 on build GRCh38
Cytoband
1p22.1
Assay ID Hs06513829_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

24

Gene Name:

ATP binding cassette subfamily A member 4

Gene Aliases:

ABC10, ABCR, ARMD2, CORD3, FFM, RMP, RP19, STGD, STGD1

Location:

Chr.1:93992837-94121149 on Build GRCh38

Assay Gene Location:

Within Intron 38
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ABCA4 NM_000350.2 NP_000341.2
AB210040.1
AF000148.1 AAC23915.1
AF001945.1 AAC05632.1
AK296896.1
AK301733.1
AK310492.1
CR627391.1 CAH10486.1
U66691.1
U88667.1 AAC51144.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv516137 Chr.1:93938682 - 94038989 on Build GRCh38 Loss ABCA4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
phospholipid transfer to membrane
transport
lipid transport
visual perception
phototransduction, visible light
phospholipid translocation
photoreceptor cell maintenance
transmembrane transport
phospholipid-translocating ATPase activity
transporter activity
eye pigment precursor transporter activity
ATP binding
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline