This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06523932_cn
See other FLJ37453 CNV Assays ›
Gene Symbol
FLJ37453
Assay Reference Genome
Location

Chr.1:15847247 on build GRCh38
Cytoband
1p36.21
Assay ID Hs06523932_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

729614

Gene Name:

uncharacterized LOC729614

Gene Aliases:

-

Location:

Chr.1:15834215-15848147 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FLJ37453 NR_024279.1
AK124018.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1011969 Chr.1:15826990 - 15875045 on Build GRCh38 Gain+Loss SPEN FLJ37453
nsv833647 Chr.1:15794042 - 15952334 on Build GRCh38 Loss ZBTB17 SPEN UQCRHL FLJ37453
dgv37n100 Chr.1:15823408 - 15924038 on Build GRCh38 Gain SPEN FLJ37453
esv2764241 Chr.1:15827002 - 15855150 on Build GRCh38 Loss SPEN FLJ37453
nsv3409 Chr.1:15809988 - 15852694 on Build GRCh38 Deletion SPEN FLJ37453
dgv38n100 Chr.1:15839166 - 15924038 on Build GRCh38 Gain SPEN FLJ37453

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline