This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06536693_cn
See other SCP2 CNV Assays ›
Gene Symbol
SCP2
Assay Reference Genome
Location

Chr.1:52929026 on build GRCh38
Cytoband
1p32.3
Assay ID Hs06536693_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

6342

Gene Name:

sterol carrier protein 2

Gene Aliases:

NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX

Location:

Chr.1:52927229-53051617 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SCP2 NM_001007098.2 NP_001007099.1
NM_001193599.1 NP_001180528.1
NM_001193600.1 NP_001180529.1
NM_001193617.1 NP_001180546.1
NM_002979.4 NP_002970.2
XM_005271103.4 XP_005271160.1
XM_011541935.2 XP_011540237.1
XM_017002045.1 XP_016857534.1
AB208789.1 BAD92026.1
AK294631.1
AK295214.1
AK308105.1
AK312856.1
BC067108.1 AAH67108.1
DC330171.1
M75883.1 AAA03557.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1001535 Chr.1:52620368 - 53346539 on Build GRCh38 Gain LOC105378732 ZYG11A COA7 MIR1273F FAM159A ZYG11B LOC100507564 PODN CPT2 ECHDC2 C1orf123 MIR1273G LRP8 MAGOH MIR5095 SCP2 SLC1A7

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

acyltransferase transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

bile acid biosynthetic process
peroxisome organization
sterol transport
positive regulation of intracellular cholesterol transport
fatty acid beta-oxidation using acyl-CoA oxidase
alpha-linolenic acid metabolic process
lipid hydroperoxide transport
fatty-acyl-CoA binding
receptor binding
protein binding
sterol transporter activity
cholesterol binding
propanoyl-CoA C-acyltransferase activity
long-chain fatty acyl-CoA binding
propionyl-CoA C2-trimethyltridecanoyltransferase activity
oleic acid binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bm6jx:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0