This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06538026_cn
See other NSUN4 CNV Assays ›
Gene Symbol
NSUN4
Assay Reference Genome
Location

Chr.1:46361253 on build GRCh38
Cytoband
1p33
Assay ID Hs06538026_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

387338

Gene Name:

NOP2/Sun RNA methyltransferase family member 4

Gene Aliases:

SHTAP

Location:

Chr.1:46340177-46365152 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NSUN4 NM_001256127.1 NP_001243056.1
NM_001256128.1 NP_001243057.1
NM_199044.3 NP_950245.2
NR_045789.1
NR_045790.1
NR_045791.1
AK021577.1 BAB13847.1
AK057420.1
AK097524.1
AK128066.1 BAC87258.1
AK291741.1
AK292353.1
AK295003.1
BC014441.1 AAH14441.1
BX648592.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv829748 Chr.1:46258836 - 46410450 on Build GRCh38 Loss UQCRH LRRC41 FAAH RAD54L NSUN4
esv2761743 Chr.1:45874186 - 46561071 on Build GRCh38 Gain DMBX1 PIK3R3 POMGNT1 LRRC41 FAAH NSUN4 MAST2 LURAP1 LINC01398 FAAHP1 TSPAN1 UQCRH RAD54L MKNK1-AS1 LOC101929626 KNCN MKNK1
nsv546162 Chr.1:46341925 - 46405089 on Build GRCh38 Loss FAAH NSUN4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

rRNA methylation
mature ribosome assembly
positive regulation of mitochondrial translation
protein binding
methyltransferase activity
small ribosomal subunit rRNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline