This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06578070_cn
See other AADACL4 CNV Assays ›
Gene Symbol
AADACL4
Assay Reference Genome
Location

Chr.1:12650208 on build GRCh38
Cytoband
1p36.21
Assay ID Hs06578070_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

343066

Gene Name:

arylacetamide deacetylase like 4

Gene Aliases:

-

Location:

Chr.1:12644547-12667086 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AADACL4 NM_001013630.1 NP_001013652.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3585236 Chr.1:12642968 - 12652029 on Build GRCh38 Loss AADACL4
nsv1011540 Chr.1:12601623 - 12832443 on Build GRCh38 Loss PRAMEF12 C1orf158 DHRS3 AADACL3 AADACL4 PRAMEF1 PRAMEF11

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

deacetylase

Gene Ontology Categories:

Function(s) Process(es)

catabolic process
hydrolase activity
carboxylic ester hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0