This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06585082_cn
See other SLC2A7 CNV Assays ›
Gene Symbol
SLC2A7
Assay Reference Genome
Location

Chr.1:9016149 on build GRCh38
Cytoband
1p36.23
Assay ID Hs06585082_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

155184

Gene Name:

solute carrier family 2 member 7

Gene Aliases:

GLUT7

Location:

Chr.1:8992534-9026384 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC2A7 NM_207420.2 NP_997303.2
XM_011540824.2 XP_011539126.1
XM_011540825.2 XP_011539127.1
AY571960.2

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv545345 Chr.1:8831066 - 9035551 on Build GRCh38 Gain ENO1-AS1 SLC2A7 MIR6728 ENO1 CA6
nsv1004025 Chr.1:9000983 - 9069086 on Build GRCh38 Gain SLC2A7 SLC2A5
nsv545346 Chr.1:8998487 - 9020907 on Build GRCh38 Gain SLC2A7
dgv15n100 Chr.1:8992095 - 9056684 on Build GRCh38 Loss SLC2A7 SLC2A5

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate transmembrane transport
sugar transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline