This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06586900_cn
See other ENO1 CNV Assays ›
Gene Symbol
ENO1
Assay Reference Genome
Location

Chr.1:8872960 on build GRCh38
Cytoband
1p36.23
Assay ID Hs06586900_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2023

Gene Name:

enolase 1

Gene Aliases:

ENO1L1, HEL-S-17, MPB1, NNE, PPH

Location:

Chr.1:8861000-8879092 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ENO1 NM_001428.3 NP_001419.1
XM_006710433.2 XP_006710496.1
AK098420.1
AK222517.1 BAD96237.1
AK223192.1 BAD96912.1
AK309177.1
AK309291.1
AK315417.1
BC001810.2 AAH01810.1
BC004458.1 AAH04458.1
BC009912.2 AAH09912.1
BC011130.1 AAH11130.1
BC015641.2 AAH15641.1
BC022545.1 AAH22545.1
BC027725.2 AAH27725.1
BC050642.2 AAH50642.1
BT007163.1 AAP35827.1
BX537400.1 CAD97642.1
GQ472226.1
M14328.1 AAA52387.1
X84907.1 CAA59331.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv545345 Chr.1:8831066 - 9035551 on Build GRCh38 Gain ENO1-AS1 CA6 MIR6728 ENO1 SLC2A7
nsv1001865 Chr.1:8843872 - 8873183 on Build GRCh38 Gain MIR6728 ENO1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
gluconeogenesis
transcription, DNA-templated
response to virus
negative regulation of cell growth
negative regulation of transcription, DNA-templated
positive regulation of binding
canonical glycolysis
cell-cell adhesion
magnesium ion binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
phosphopyruvate hydratase activity
protein binding
poly(A) RNA binding
protein heterodimerization activity
GTPase binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hlj6l:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0