This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06596105_cn
See other ARID1A CNV Assays ›
Gene Symbol
ARID1A
Assay Reference Genome
Location

Chr.1:26717580 on build GRCh38
Cytoband
1p36.11
Assay ID Hs06596105_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8289

Gene Name:

AT-rich interaction domain 1A

Gene Aliases:

B120, BAF250, BAF250a, BM029, C1orf4, CSS2, ELD, MRD14, OSA1, P270, SMARCF1, hELD, hOSA1

Location:

Chr.1:26696031-26782110 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ARID1A NM_006015.4 NP_006006.3
NM_139135.2 NP_624361.1
AB210022.1 BAE06104.1
AF219114.1 AAG17549.2
AF231056.1 AAG33967.1
AF265208.1 AAF75765.1
AF521670.1 AAN03446.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1000012 Chr.1:26714969 - 27141528 on Build GRCh38 Gain KDF1 NUDC GPATCH3 NR0B2 ARID1A FAM46B TRNP1 ZDHHC18 SFN SLC9A1 PIGV GPN2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural tube closure
cardiac chamber development
optic cup formation involved in camera-type eye development
nucleosome disassembly
chromatin remodeling
maintenance of chromatin silencing
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
covalent chromatin modification
intracellular estrogen receptor signaling pathway
androgen receptor signaling pathway
forebrain development
nucleosome mobilization
glucocorticoid receptor signaling pathway
ATP-dependent chromatin remodeling
positive regulation of transcription, DNA-templated
chromatin-mediated maintenance of transcription
cardiac muscle cell differentiation
placenta blood vessel development
toxin transport
DNA binding
transcription coactivator activity
protein binding
ligand-dependent nuclear receptor binding
nucleosome binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline