This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06651838_cn
See other NFKBIZ CNV Assays ›
Gene Symbol
NFKBIZ
Assay Reference Genome
Location

Chr.3:101830159 on build GRCh38
Cytoband
3q12.3
Assay ID Hs06651838_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

64332

Gene Name:

NFKB inhibitor zeta

Gene Aliases:

IKBZ, INAP, MAIL

Location:

Chr.3:101827990-101861025 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NFKBIZ NM_001005474.2 NP_001005474.1
AB074898.1
AF548362.1 AAN40698.1
AK054787.1
BM050734.1
DA541852.1
DQ224339.1 ABB02425.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv953238 Chr.3:101825757 - 101845056 on Build GRCh38 Duplication NXPE3 NFKBIZ

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
inflammatory response
transcription cofactor activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-bxd7r:80/100.66.79.78:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline