Assay Details
Target Gene Details
Entrez Gene ID: | 58528 |
Gene Name: | Ras related GTP binding D |
Gene Aliases: |
RAGD, bA11D8.2.1 |
Location: |
Chr.6:89363509-89412470 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAGD | NM_021244.4 | NP_067067.1 | ||
| XM_005248755.4 | XP_005248812.1 | |||
| XM_011536022.2 | XP_011534324.1 | |||
| AF272036.1 | AAG32663.1 | |||
| AK289799.1 | ||||
| AL137502.1 | CAB70775.2 | |||
| BC003088.1 | AAH03088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
| nsv1027007 | Chr.6:89099218 - 89628146 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map